Assay Details
Gene Symbol: LOC100280491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153411.1 | NP_001146883.1 | 821 | 67 | ||
GenBank mRNA | EU952042.1 | - | 821 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
