Assay Details
Gene Symbol: LOC100281203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154122.1 | NP_001147594.1 | 552 | 67 | ||
GenBank mRNA | BT037540.1 | - | 318 | 67 | ||
BT041844.1 | - | 639 | 67 | |||
EU955911.1 | - | 552 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
