Assay Details
Gene Symbol: LOC100281203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154122.1 | NP_001147594.1 | 773 | 71 | ||
GenBank mRNA | BT037540.1 | - | 539 | 71 | ||
BT041844.1 | - | 860 | 71 | |||
EU955911.1 | - | 773 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
