Assay Details
Gene Symbol: LOC100282749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155655.1 | NP_001149127.1 | 552 | 67 | ||
GenBank mRNA | BT039024.1 | - | 552 | 67 | ||
BT054021.1 | - | 391 | 67 | |||
BT067680.1 | - | 529 | 67 | |||
EU962180.1 | - | 552 | 67 |
