Assay Details
Gene Symbol: LOC100283062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155964.1 | NP_001149436.1 | 153 | 64 | ||
GenBank mRNA | BT040601.1 | - | 64 | |||
BT040953.1 | - | 64 | ||||
BT042202.1 | - | 64 | ||||
EU963324.1 | - | 153 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
