Assay Details
Gene Symbol: LOC100283445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156346.1 | NP_001149818.1 | 694 | 66 | ||
GenBank mRNA | BT065513.1 | - | 643 | 66 | ||
EU964781.1 | - | 694 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
