Assay Details
Gene Symbol: LOC100284035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156933.1 | NP_001150405.1 | 505 | 70 | ||
GenBank mRNA | EU966849.1 | - | 505 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
