Assay Details
Gene Symbol: LOC100284112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157009.1 | NP_001150481.1 | 398 | 69 | ||
GenBank mRNA | BT061701.1 | - | 328 | 69 | ||
EU967127.1 | - | 398 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
