Assay Details
| Gene Symbol: LOC100284112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157009.1 | NP_001150481.1 | 1319 | 64 | ||
| GenBank mRNA | EU967127.1 | - | 1319 | 64 | ||
More Information
Set Membership: |
|

LOC100284112
Genomic Map