Assay Details
Gene Symbol: LOC100284499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157394.1 | NP_001150866.1 | 1186 | 97 | ||
GenBank mRNA | EU968583.1 | - | 1186 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
