Assay Details
Gene Symbol: LOC100285205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158099.1 | NP_001151571.1 | 1040 | 74 | ||
GenBank mRNA | EU971271.1 | - | 1040 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
