Assay Details
Gene Symbol: LOC100285205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158099.1 | NP_001151571.1 | 1963 | 87 | ||
GenBank mRNA | EU971271.1 | - | 1963 | 87 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100285205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158099.1 | NP_001151571.1 | 1963 | 87 | ||
GenBank mRNA | EU971271.1 | - | 1963 | 87 |