Assay Details
| Gene Symbol: LOC100285310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158203.1 | NP_001151675.1 | 142 | 55 | ||
| GenBank mRNA | BT088061.1 | - | 133 | 55 | ||
| EU971703.1 | - | 142 | 55 | |||
More Information
Set Membership: |
|

LOC100285310
Genomic Map