Assay Details
Gene Symbol: rrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111406.1 | NP_001104876.1 | 183 | 95 | ||
GenBank mRNA | AF007793.1 | - | 183 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
