Assay Details
Gene Symbol: rrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111406.1 | NP_001104876.1 | 806 | 80 | ||
GenBank mRNA | AF007793.1 | - | 806 | 80 | ||
AF250050.1 | - | 826 | 80 | |||
X98923.1 | - | 188 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
