Assay Details
| Gene Symbol: hda108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111431.1 | NP_001104901.1 | 1223 | 84 | ||
| GenBank mRNA | AF045473.1 | - | 1223 | 84 | ||
| AF440226.1 | - | 1157 | 84 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

hda108| Gene Symbol: hda108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111431.1 | NP_001104901.1 | 1223 | 84 | ||
| GenBank mRNA | AF045473.1 | - | 1223 | 84 | ||
| AF440226.1 | - | 1157 | 84 | |||