Assay Details
| Gene Symbol: csu134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111477.1 | NP_001104947.1 | 1264 | 70 | ||
| GenBank mRNA | AF174486.1 | - | 1264 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

csu134| Gene Symbol: csu134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111477.1 | NP_001104947.1 | 1264 | 70 | ||
| GenBank mRNA | AF174486.1 | - | 1264 | 70 | ||
Set Membership: |
|