Assay Details
Gene Symbol: phb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111497.1 | NP_001104967.1 | 945 | 80 | ||
GenBank mRNA | AF236369.1 | - | 945 | 80 | ||
AY106200.1 | - | 945 | 80 | |||
BT087806.1 | - | 873 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
