Assay Details
Gene Symbol: ckb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111504.1 | NP_001104974.1 | 900 | 97 | ||
GenBank mRNA | AF239816.1 | - | 900 | 97 | ||
BT040828.1 | - | 875 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
