Assay Details
| Gene Symbol: LOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111533.1 | NP_001105003.1 | 298 | 74 | ||
| GenBank mRNA | AF271894.1 | - | 218 | 74 | ||
| BT067411.1 | - | 256 | 74 | |||
| DQ335760.1 | - | 298 | 74 | |||
More Information
Set Membership: |
|

LOX
Genomic Map