Assay Details
| Gene Symbol: akin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111535.1 | NP_001105005.1 | 1040 | 99 | ||
| GenBank mRNA | AF276085.1 | - | 1040 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

akin1| Gene Symbol: akin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111535.1 | NP_001105005.1 | 1040 | 99 | ||
| GenBank mRNA | AF276085.1 | - | 1040 | 99 | ||
Set Membership: |
|