Assay Details
Gene Symbol: pip2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111557.1 | NP_001105027.1 | 841 | 72 | ||
GenBank mRNA | AF326495.1 | - | 841 | 72 | ||
BT039930.1 | - | 820 | 72 | |||
EU964089.1 | - | 846 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
