Assay Details
| Gene Symbol: pao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111636.1 | NP_001105106.1 | 202 | 67 | ||
| GenBank mRNA | AJ002204.1 | - | 202 | 67 | ||
| AY107888.1 | - | 209 | 67 | |||
More Information
Set Membership: |
|

pao1
Genomic Map