Assay Details
| Gene Symbol: mlo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111698.1 | NP_001105168.1 | 1018 | 72 | ||
| GenBank mRNA | AY029313.1 | - | 1018 | 72 | ||
| AY105309.1 | - | 1018 | 72 | |||
| BT063312.1 | - | 1049 | 72 | |||

mlo2
Genomic Map