Assay Details
Gene Symbol: mlo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111699.1 | NP_001105169.1 | 934 | 81 | ||
GenBank mRNA | AY029315.1 | - | 934 | 81 | ||
AY103929.1 | - | 936 | 81 | |||
BT035073.1 | - | 1199 | 81 | |||
BT066636.1 | - | 943 | 81 |
