Assay Details
| Gene Symbol: mlo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111700.1 | NP_001105170.1 | 1050 | 65 | ||
| GenBank mRNA | AY029317.1 | - | 1050 | 65 | ||
| AY108340.1 | - | 1050 | 65 | |||
| BT062352.1 | - | 1152 | 65 | |||
More Information
Set Membership: |
|

mlo6
Genomic Map