Assay Details
Gene Symbol: sus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111853.1 | NP_001105323.1 | 1728 | 60 | ||
GenBank mRNA | AY103630.1 | - | 1864 | 60 | ||
L22296.1 | - | 1728 | 60 |
