Assay Details
Gene Symbol: sus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111853.1 | NP_001105323.1 | 134 | 66 | ||
GenBank mRNA | AY103630.1 | - | 269 | 66 | ||
L22296.1 | - | 134 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
