Assay Details
Gene Symbol: sus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111853.1 | NP_001105323.1 | 270 | 80 | ||
GenBank mRNA | AY103630.1 | - | 405 | 80 | ||
L22296.1 | - | 270 | 80 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: sus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111853.1 | NP_001105323.1 | 270 | 80 | ||
GenBank mRNA | AY103630.1 | - | 405 | 80 | ||
L22296.1 | - | 270 | 80 |