Assay Details
Gene Symbol: sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111865.1 | NP_001105335.1 | 308 | 78 | ||
GenBank mRNA | EU958371.1 | - | 332 | 78 | ||
EU958837.1 | - | 349 | 78 | |||
M15175.1 | - | 308 | 78 | |||
M54936.1 | - | 308 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
