Assay Details
| Gene Symbol: cyc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111892.1 | NP_001105362.1 | 977 | 75 | ||
| GenBank mRNA | AY111119.1 | - | 977 | 75 | ||
| EU961361.1 | - | 1098 | 75 | |||
| U10076.1 | - | 977 | 75 | |||
More Information
Set Membership: |
|

cyc3
Genomic Map