Assay Details
Gene Symbol: ACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111903.1 | NP_001105373.1 | 3816 | 105 | ||
GenBank mRNA | AY312172.1 | - | 3787 | 105 | ||
U19183.1 | - | 3816 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
