Assay Details
Gene Symbol: ACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111903.1 | NP_001105373.1 | 1033 | 71 | ||
GenBank mRNA | AY312171.1 | - | 1004 | 71 | ||
AY312172.1 | - | 1004 | 71 | |||
U19183.1 | - | 1033 | 71 |
