Assay Details
| Gene Symbol: pds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111911.1 | NP_001105381.1 | 2004 | 78 | ||
| GenBank mRNA | AY109597.1 | - | 2005 | 78 | ||
| L39266.1 | - | 78 | ||||
| U37285.1 | - | 2004 | 78 | |||
More Information
Set Membership: |
|

pds1
Genomic Map