Assay Details
| Gene Symbol: dnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112042.1 | NP_001105512.1 | 251 | 81 | ||
| GenBank mRNA | AF288075.1 | - | 251 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dnp1| Gene Symbol: dnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112042.1 | NP_001105512.1 | 251 | 81 | ||
| GenBank mRNA | AF288075.1 | - | 251 | 81 | ||
Set Membership: |
|