Assay Details
| Gene Symbol: rps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112074.1 | NP_001105544.1 | 312 | 69 | ||
| GenBank mRNA | EU959790.1 | - | 338 | 69 | ||
| U92045.1 | - | 312 | 69 | |||
More Information
Set Membership: |
|

rps6
Genomic Map