Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112112.1 | NP_001105582.1 | 660 | 69 | ||
| GenBank mRNA | AY156979.1 | - | 660 | 69 | ||
| BT064118.1 | - | 713 | 69 | |||
| BT064143.1 | - | 601 | 69 | |||
| BT067700.1 | - | 629 | 69 | |||
| EU966640.1 | - | 679 | 69 | |||
More Information
Set Membership: |
|

NIT1
Genomic Map