Assay Details
| Gene Symbol: pip2-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112146.1 | NP_001105616.1 | 668 | 67 | ||
| GenBank mRNA | AF130975.1 | - | 668 | 67 | ||
| AY243802.1 | - | 613 | 67 | |||
More Information
Set Membership: |
|

pip2-5
Genomic Map