Assay Details
Gene Symbol: mlo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112190.1 | NP_001105660.1 | 571 | 68 | ||
GenBank mRNA | AY029312.2 | - | 571 | 68 | ||
BT036101.1 | - | 676 | 68 |
