Assay Details
Gene Symbol: mlo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112190.1 | NP_001105660.1 | 887 | 83 | ||
GenBank mRNA | AY029312.2 | - | 887 | 83 | ||
AY110401.1 | - | 671 | 83 | |||
BT036101.1 | - | 992 | 83 |
