Assay Details
Gene Symbol: mlo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112191.1 | NP_001105661.1 | 163 | 88 | ||
GenBank mRNA | AY029318.1 | - | 163 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
