Assay Details
| Gene Symbol: mlo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112191.1 | NP_001105661.1 | 496 | 83 | ||
| GenBank mRNA | AY029318.1 | - | 496 | 83 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlo7| Gene Symbol: mlo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112191.1 | NP_001105661.1 | 496 | 83 | ||
| GenBank mRNA | AY029318.1 | - | 496 | 83 | ||