Assay Details
| Gene Symbol: fea2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112192.1 | NP_001105662.1 | 1749 | 102 | ||
| GenBank mRNA | AY055124.1 | - | 1749 | 102 | ||
| BT042749.2 | - | 1785 | 102 | |||
More Information
Set Membership: |
|

fea2
Genomic Map