Assay Details
| Gene Symbol: eif3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112256.1 | NP_001105726.1 | 1527 | 105 | ||
| GenBank mRNA | AF073329.1 | - | 1527 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eif3| Gene Symbol: eif3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112256.1 | NP_001105726.1 | 1527 | 105 | ||
| GenBank mRNA | AF073329.1 | - | 1527 | 105 | ||
Set Membership: |
|