Assay Details
| Gene Symbol: rop5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112267.1 | NP_001105737.1 | 374 | 69 | ||
| GenBank mRNA | AJ278666.1 | - | 374 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rop5| Gene Symbol: rop5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112267.1 | NP_001105737.1 | 374 | 69 | ||
| GenBank mRNA | AJ278666.1 | - | 374 | 69 | ||
Set Membership: |
|