Assay Details
Gene Symbol: rop5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112267.1 | NP_001105737.1 | 438 | 64 | ||
GenBank mRNA | AJ278666.1 | - | 438 | 64 |
Gene Symbol: LOC100381374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174224.1 | NP_001167695.1 | 629 | 64 | ||
GenBank mRNA | EU970190.1 | - | 629 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
