Assay Details
| Gene Symbol: pdi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112288.1 | NP_001105758.1 | 676 | 90 | ||
| GenBank mRNA | AY739288.1 | - | 676 | 90 | ||
| BT063570.1 | - | 674 | 90 | |||
More Information
Set Membership: |
|

pdi6
Genomic Map