Assay Details
Gene Symbol: AGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112324.1 | NP_001105794.1 | 2055 | 80 | ||
GenBank mRNA | AF497511.2 | - | 2055 | 80 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112324.1 | NP_001105794.1 | 2055 | 80 | ||
GenBank mRNA | AF497511.2 | - | 2055 | 80 |