Assay Details
Gene Symbol: si603015d01b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112344.1 | NP_001105814.1 | 912 | 90 | ||
GenBank mRNA | DQ436921.1 | - | 912 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
