Assay Details
Gene Symbol: crp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112409.1 | NP_001105879.1 | 1940 | 94 | ||
GenBank mRNA | AF073522.1 | - | 1940 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
